Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290311.1 | NP_001277240.1 | 3 | 1386 | ||
NM_001290313.1 | NP_001277242.1 | 3 | 1386 | |||
NM_029361.4 | NP_083637.2 | 3 | 1386 | |||
GenBank mRNA | AK147550.1 | - | 3 | 1073 |