Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029393.1 | NP_083669.1 | 5 | 1139 | ||
GenBank mRNA | AK009986.1 | - | 4 | 484 | ||
AK010165.1 | - | 5 | 1162 | |||
AK053678.1 | - | 5 | 885 | |||
BC129847.1 | - | 5 | 1139 | 40107361 | ||
BC129848.1 | - | 5 | 1139 | 40107363 | ||
GU046702.1 | - | 4 | 1203 |