Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019276.3 | NP_062149.1 | 2 | 453 | ||
GenBank mRNA | L21698.1 | - | 2 | 500 |