Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290811.1 | NP_001277740.1 | 10 | 1160 | ||
NM_001290812.1 | NP_001277741.1 | 9 | 1054 | |||
NM_029475.2 | NP_083751.1 | 10 | 1052 | |||
GenBank mRNA | AK039677.1 | - | 9 | 1045 | ||
AK049697.1 | - | 10 | 1153 | |||
BC050879.1 | - | 10 | 1004 | 5687835 | ||
BC052048.1 | - | 10 | 1004 | 5687835 |