Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162956.1 | NP_001156428.1 | 16 | 2336 | ||
GenBank mRNA | AK080572.1 | - | 2 | 530 | ||
BC042726.1 | - | 6 | 1166 |