Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019285.2 | NP_062158.2 | 14, 15 | 1852 | ||
GenBank mRNA | BC081813.1 | - | 14, 15 | 1852 | 7125413 | |
FQ223268.1 | - | 13, 14 | 1980 | |||
FQ223451.1 | - | 13, 14 | 1980 | |||
M80633.1 | - | 14, 15 | 1818 |