Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc18b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183116.2 | NP_898939.2 | 3 | 423 | ||
GenBank mRNA | AK183134.1 | - | 3 | 333 | ||
CK625631.1 | - | 3 | 353 |