Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc18b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183116.2 | NP_898939.2 | 11, 12 | 1323 | ||
GenBank mRNA | BC068251.1 | - | 6, 7 | 588 | ||
BM949736.1 | - | 5, 6 | 507 |