Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190156.1 | NP_001177085.1 | 8 | 1204 | ||
NM_029655.3 | NP_083931.2 | 9 | 1357 | |||
GenBank mRNA | AK011015.1 | - | 9 | 1332 | ||
AK038971.1 | - | 9 | 1358 | |||
AK052487.1 | - | 9 | 1314 | |||
AK170966.1 | - | 8 | 1151 | |||
BC087942.1 | - | 9 | 1215 | 30303462 |