Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700113H08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029685.1 | NP_083961.1 | 6 | 1306 | ||
GenBank mRNA | AK007195.1 | - | 2 | 777 |