Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESRRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282450.1 | NP_001269379.1 | 6 | 1106 | ||
NM_001282451.1 | NP_001269380.1 | 6 | 1090 | |||
NM_004451.4 | NP_004442.3 | 6 | 1093 | |||
XM_017017313.1 | XP_016872802.1 | 5 | 1597 | |||
GenBank mRNA | BC007915.2 | - | 5 | 563 | ||
BC033701.1 | - | 4 | 502 | |||
BC063795.1 | - | 6 | 1043 | 6527166 | ||
BC092470.1 | - | 6 | 1106 | 30336569 | ||
BC093720.1 | - | 5 | 894 | 7939565 | ||
BC093722.1 | - | 5 | 894 | 7939567 | ||
HQ692867.1 | - | 5 | 872 | |||
L38487.1 | - | 6 | 1046 | |||
X51416.1 | - | 1226 |