Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gje1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029722.1 | NP_083998.1 | 3 | 765 | ||
GenBank mRNA | AK018698.1 | - | 3 | 766 |