Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133972.2 | NP_598733.2 | 4 | 926 | ||
GenBank mRNA | AK010815.1 | - | 4 | 954 | ||
AK049351.1 | - | 4 | 642 | |||
AK080609.1 | - | 4 | 631 | |||
AK081550.1 | - | 4 | 971 | |||
BC005754.1 | - | 1 | 135 | |||
BC043070.1 | - | 4 | 926 | 6403587 | ||
BC048396.1 | - | 4 | 887 | |||
BC055779.1 | - | 4 | 926 | 6403587 |