Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calcoco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271018.1 | NP_001257947.1 | 6 | 1643 | ||
GenBank mRNA | BB730060.1 | - | 1 | 86 |