Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufaf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085493.2 | NP_001078962.1 | 9 | 919 | ||
GenBank mRNA | AK009547.1 | - | 9 | 900 | ||
BC048539.1 | - | 3 | 169 |