Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpped2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143683.1 | NP_001137155.1 | 7 | 1825 | ||
NM_029837.1 | NP_084113.1 | 7 | 1538 | |||
GenBank mRNA | AK164559.1 | - | 1 | 325 | ||
AK165186.1 | - | 7 | 2022 |