Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C330018D20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029909.1 | NP_084185.1 | 5 | 888 | ||
GenBank mRNA | AK021208.1 | - | 5 | 866 | ||
AK140255.1 | - | 5 | 869 |