Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C330018D20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029909.1 | NP_084185.1 | 5 | 378 | ||
GenBank mRNA | AK021208.1 | - | 5 | 355 | ||
AK140255.1 | - | 5 | 359 | |||
BC038160.1 | - | 4 | 322 | |||
BC116244.1 | - | 4 | 344 | |||
BC116245.1 | - | 4 | 344 | |||
BC127172.1 | - | 4 | 344 | |||
CD561591.1 | - | 5 | 378 |