Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jrkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033181.1 | NP_001028353.1 | 1 | 2592 | ||
GenBank mRNA | AK021123.1 | - | 1 | 773 | ||
AK079877.1 | - | 1 | 1717 | |||
BB210598.2 | - | 1 | 297 | |||
BC138472.1 | - | 1 | 2593 | 8861492 |