Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029936.2 | NP_084212.2 | 18 | 2762 | ||
GenBank mRNA | AK019495.1 | - | 18 | 2741 | ||
AK166787.1 | - | 6 | 961 | |||
BC049261.1 | - | 18 | 2703 | |||
BC080729.1 | - | 18 | 2730 | 6504816 |