Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029936.2 | NP_084212.2 | 5, 6 | 767 | ||
GenBank mRNA | AK019495.1 | - | 5, 6 | 747 | ||
AK045032.1 | - | 5, 6 | 732 | |||
AK052540.1 | - | 5, 6 | 747 | |||
BC023303.1 | - | 5, 6 | 715 | |||
BC049261.1 | - | 5, 6 | 708 | |||
BC055481.1 | - | 4, 5 | 485 | |||
BC080729.1 | - | 5, 6 | 735 | 6504816 |