Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acbd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030063.2 | NP_084339.1 | 1 | 30 | ||
GenBank mRNA | AK020348.1 | - | 1 | 31 |