Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081335.2 | NP_001074804.2 | 31, 32 | 6439 | ||
GenBank mRNA | AK051474.1 | - | 8, 9 | 1667 | ||
AK122351.1 | - | 28, 29 | 5429 | |||
AK147675.1 | - | 29, 30 | 7029 | |||
AK162968.1 | - | 10, 11 | 1900 |