Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162925.1 | NP_001156397.1 | 6, 7 | 823 | ||
NM_030094.1 | NP_084370.1 | 6, 7 | 772 | |||
GenBank mRNA | AK019109.1 | - | 6, 7 | 777 | ||
AK134929.1 | - | 6, 7 | 760 | |||
AK141256.1 | - | 6, 7 | 760 | |||
BC115559.2 | - | 6, 7 | 808 | 40101853 |