Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033186.3 | NP_001028358.1 | 12, 13 | 1198 | ||
NM_001177898.1 | NP_001171369.1 | 10, 11 | 1041 | |||
NM_001177899.1 | NP_001171370.1 | 11, 12 | 1150 | |||
GenBank mRNA | AK018920.1 | - | 3, 4 | 682 | ||
AK137942.1 | - | 12, 13 | 1166 | |||
AK138030.1 | - | 10, 11 | 1024 | |||
AK169825.1 | - | 11, 12 | 1115 | |||
BC145402.1 | - | 11, 12 | 1079 | 9053960 | ||
BC150764.1 | - | 11, 12 | 1079 | 9087675 | ||
BC171961.1 | - | 11, 12 | 1079 | 9054247 |