Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micu3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030110.1 | NP_084386.1 | 8, 9 | 865 | ||
GenBank mRNA | AK086433.1 | - | 1, 2 | 2325 | ||
BC125419.1 | - | 7, 8 | 459 | |||
BC125445.1 | - | 7, 8 | 459 |