Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Filip1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040397.4 | NP_001035487.2 | 5 | 3555 | ||
NM_001177871.1 | NP_001171342.1 | 2 | 2862 | |||
GenBank mRNA | AK019472.2 | - | 2 | 2863 | ||
AK132319.1 | - | 2 | 2863 | |||
BC037751.1 | - | 1 | 599 | |||
BC062166.1 | - | 2 | 2845 | |||
BC098507.1 | - | 1 | 2699 | |||
EU623548.1 | - | 1 | 2674 | |||
EU704258.1 | - | 5 | 3402 |