Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930562C15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030192.1 | NP_084468.1 | 1 | 414 | ||
GenBank mRNA | AK133365.1 | - | 1 | 414 |