Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159275.1 | NP_001152747.1 | 1 | 1153 | ||
GenBank mRNA | AF332006.1 | - | 1 | 1011 | ||
AK077159.1 | - | 1 | 1154 | |||
AY583456.1 | - | 1 | 1138 | |||
BC114348.1 | - | 1 | 995 | 40051484 | ||
BC125010.1 | - | 1 | 995 | 40051483 |