Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019365.3 | NP_062238.1 | 3 | 748 | ||
XM_006249796.2 | XP_006249858.1 | 3 | 1609 | |||
XM_008769494.1 | XP_008767716.1 | 2 | 427 | |||
GenBank mRNA | AF002251.1 | - | 3 | 748 |