Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itih4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019369.2 | NP_062242.2 | 2 | 247 | ||
GenBank mRNA | BC089806.1 | - | 2 | 247 | 7326076 | |
Y11283.1 | - | 2 | 247 |