Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019905.1 | NP_063970.1 | 3 | 160 | ||
GenBank mRNA | BC059136.1 | - | 3 | 179 | 6920034 | |
FQ225322.1 | - | 3 | 176 | |||
L13039.1 | - | 2 | 131 | |||
S73559.1 | - | 3 | 162 | |||
X66871.1 | - | 3 | 160 |