Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020092.1 | NP_064477.1 | 3 | 315 | ||
XM_006231892.2 | XP_006231954.1 | 5 | 834 | |||
XM_006231894.2 | XP_006231956.1 | 4 | 5243 | |||
GenBank mRNA | AF234765.1 | - | 3 | 315 |