Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019328.3 | NP_062201.2 | 2, 3 | 972 | ||
XM_006234201.2 | XP_006234263.1 | 3, 4 | 1190 | |||
XM_008761861.1 | XP_008760083.1 | 2, 3 | 1106 | |||
GenBank mRNA | DQ092629.1 | - | 2, 3 | 903 | ||
DQ092630.1 | - | 2, 3 | 739 | |||
DQ092631.1 | - | 2, 3 | 917 | |||
DQ092632.1 | - | 2, 3 | 907 | |||
DQ092633.1 | - | 2, 3 | 739 | |||
DQ092634.1 | - | 2, 3 | 739 | |||
FQ222109.1 | - | 3, 4 | 1211 | |||
L08595.1 | - | 2, 3 | 972 | |||
U01146.1 | - | 2, 3 | 903 | |||
U72345.1 | - | 3, 4 | 1196 |