Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019344.2 | NP_062217.1 | 6 | 683 | ||
XM_006250015.2 | XP_006250077.1 | 5 | 554 | |||
XM_006250016.2 | XP_006250078.1 | 6 | 498 | |||
XM_006250017.2 | XP_006250079.1 | 6 | 848 | |||
XM_008769675.1 | XP_008767897.1 | 6 | 849 | |||
XM_008769676.1 | XP_008767898.1 | 6 | 1014 | |||
GenBank mRNA | AB006013.1 | - | 5 | 592 | ||
BC089064.1 | - | 6 | 683 | 7317138 | ||
U32432.1 | - | 1 | 20 |