Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019360.2 | NP_062233.2 | 3 | 296 | ||
GenBank mRNA | BC058480.1 | - | 3 | 272 | 6922382 | |
CF976717.1 | - | 3 | 300 | |||
FQ210091.1 | - | 3 | 274 | |||
FQ210263.1 | - | 3 | 273 | |||
FQ217805.1 | - | 3 | 299 | |||
FQ221395.1 | - | 3 | 275 | |||
FQ222319.1 | - | 3 | 268 | |||
FQ222901.1 | - | 3 | 317 | |||
FQ223820.1 | - | 3 | 271 | |||
K01565.1 | - | 1 | 83 | |||
M27466.1 | - | 3 | 307 | |||
M27467.1 | - | 2 | 271 |