Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019906.1 | NP_063971.1 | 3 | 245 | ||
GenBank mRNA | L37085.1 | - | 2 | 209 | ||
U11681.1 | - | 3 | 245 |