Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019344.2 | NP_062217.1 | 7 | 904 | ||
XM_006250015.2 | XP_006250077.1 | 6 | 775 | |||
XM_006250016.2 | XP_006250078.1 | 7 | 719 | |||
XM_006250017.2 | XP_006250079.1 | 7 | 1069 | |||
XM_008769675.1 | XP_008767897.1 | 7 | 1070 | |||
XM_008769676.1 | XP_008767898.1 | 7 | 1235 | |||
GenBank mRNA | AB006013.1 | - | 6 | 813 | ||
BC089064.1 | - | 7 | 904 | 7317138 |