Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm17a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019351.1 | NP_062224.1 | 2 | 118 | ||
GenBank mRNA | AB006450.1 | - | 2 | 118 | ||
BC061982.1 | - | 2 | 165 | 5598788 |
Gene Symbol: Timm17al1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_576033.5 | XP_576033.1 | 2 | 137 |