Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019359.1 | NP_062232.1 | 5 | 636 | ||
GenBank mRNA | BC062020.1 | - | 5 | 639 | 5623281 | |
FQ228936.1 | - | 2 | 73 | |||
U06755.1 | - | 5 | 636 |