Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271899.1 | NP_001258828.1 | 3, 4 | 445 | ||
NM_001271900.1 | NP_001258829.1 | 4, 5 | 639 | |||
NM_001271948.1 | NP_001258877.1 | 3, 4 | 972 | |||
NM_181674.2 | NP_858060.2 | 3, 4 | 770 | |||
NM_181675.3 | NP_858061.2 | 3, 4 | 492 | |||
NM_181676.2 | NP_858062.1 | 2, 3 | 576 | |||
NM_181677.2 | NP_858063.1 | 4, 5 | 402 | |||
NM_181678.2 | NP_858064.1 | 4, 5 | 415 | |||
NR_073526.1 | 2, 3 | 887 | ||||
NR_073527.1 | 3, 4 | 524 | ||||
GenBank mRNA | AK056192.1 | - | 3, 4 | 452 | ||
AK289717.1 | - | 3, 4 | 421 | |||
AK294659.1 | - | 4, 5 | 548 | |||
AK295347.1 | - | 3, 4 | 416 | |||
AK314810.1 | - | 3, 4 | 769 | |||
AL540315.3 | - | 1, 2 | 105 | |||
AV726282.1 | - | 4, 5 | 321 | |||
BC031790.1 | - | 3, 4 | 660 | 4939981 | ||
BI669304.1 | - | 4, 5 | 400 | |||
BX647887.1 | - | 3, 4 | 505 | |||
DA155307.1 | - | 4, 5 | 381 | |||
DA158596.1 | - | 3, 4 | 533 | |||
DA365141.1 | - | 4, 5 | 402 | |||
DA523686.1 | - | 3, 4 | 471 | |||
DA533318.1 | - | 3, 4 | 492 | |||
M64930.1 | - | 2, 3 | 698 |