Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103691938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762404.1 | XP_008760626.1 | 2 | 108 |
Gene Symbol: LOC100359498 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002727895.4 | XP_002727941.1 | 3 | 194 | ||
XM_002727896.4 | XP_002727942.1 | 3 | 199 | |||
GenBank mRNA | FQ210925.1 | - | 3 | 163 |
Gene Symbol: Rpl35a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021264.3 | NP_067087.1 | 103 | |||
GenBank mRNA | BC061557.1 | - | 169 | 6917939 | ||
X03475.1 | - | 103 |