Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl8a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021580.1 | NP_067591.1 | 2 | 207 | ||
XM_008771599.1 | XP_008769821.1 | 2 | 143 | |||
XM_008771600.1 | XP_008769822.1 | 2 | 146 | |||
GenBank mRNA | AB009889.1 | - | 2 | 207 |
Gene Symbol: LOC103694085 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771616.1 | XP_008769838.1 | 149 | |||
XM_008771617.1 | XP_008769839.1 | 143 | ||||
XM_008771618.1 | XP_008769840.1 | 146 |