Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021589.1 | NP_067600.1 | 5 | 561 | ||
XM_006232746.2 | XP_006232808.1 | 5 | 552 | |||
GenBank mRNA | AF174586.1 | - | 2 | 155 | ||
AF174587.1 | - | 1 | 86 | |||
M85214.1 | - | 5 | 561 |