Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100911769 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762428.1 | XP_008760650.1 | 3 | 330 | ||
XM_008762429.1 | XP_008760651.1 | 3 | 327 | |||
XM_008762430.1 | XP_008760652.1 | 3 | 3772 | |||
XM_008762431.1 | XP_008760653.1 | 3 | 330 | |||
XM_008762432.1 | XP_008760654.1 | 3 | 330 | |||
XM_008762433.1 | XP_008760655.1 | 3 | 330 | |||
XM_008762434.1 | XP_008760656.1 | 3 | 330 | |||
XM_008762435.1 | XP_008760657.1 | 3 | 330 |
Gene Symbol: Epb41l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021681.2 | NP_067713.2 | 3 | 360 | ||
NM_172090.3 | NP_742087.1 | 3 | 343 | |||
XM_008762381.1 | XP_008760603.1 | 3 | 360 | |||
GenBank mRNA | AB019256.1 | - | 3 | 341 | ||
AB019257.1 | - | 3 | 360 |