Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020976.1 | NP_066125.1 | 5 | 339 | ||
GenBank mRNA | AF178086.1 | - | 5 | 339 |