Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021588.2 | NP_067599.1 | 2 | 275 | ||
GenBank mRNA | AF197916.1 | - | 2 | 248 | ||
BC070511.1 | - | 2 | 275 | 7103504 | ||
FQ214690.1 | - | 2 | 307 | |||
FQ214898.1 | - | 2 | 306 | |||
FQ214986.1 | - | 2 | 307 | |||
FQ215026.1 | - | 2 | 307 | |||
FQ215032.1 | - | 2 | 306 | |||
FQ215043.1 | - | 2 | 306 | |||
FQ215217.1 | - | 2 | 305 | |||
FQ215441.1 | - | 2 | 306 | |||
FQ215511.1 | - | 2 | 305 | |||
FQ215560.1 | - | 2 | 305 | |||
FQ215564.1 | - | 2 | 305 | |||
FQ215642.1 | - | 2 | 316 | |||
FQ215802.1 | - | 2 | 307 | |||
FQ215880.1 | - | 2 | 307 | |||
FQ215982.1 | - | 2 | 306 | |||
FQ216001.1 | - | 2 | 305 | |||
FQ216029.1 | - | 2 | 304 | |||
FQ216127.1 | - | 2 | 304 | |||
FQ216280.1 | - | 2 | 313 | |||
FQ216299.1 | - | 2 | 307 | |||
FQ216306.1 | - | 2 | 308 | |||
FQ216335.1 | - | 2 | 307 | |||
FQ216380.1 | - | 2 | 306 | |||
FQ216443.1 | - | 2 | 305 | |||
FQ216504.1 | - | 2 | 306 | |||
FQ216709.1 | - | 2 | 306 | |||
FQ216731.1 | - | 2 | 305 | |||
FQ216841.1 | - | 2 | 304 | |||
FQ217450.1 | - | 2 | 306 | |||
FQ217496.1 | - | 2 | 305 | |||
FQ218027.1 | - | 2 | 306 | |||
FQ223624.1 | - | 2 | 305 | |||
FQ223672.1 | - | 2 | 302 | |||
FQ223774.1 | - | 2 | 307 | |||
FQ223938.1 | - | 2 | 307 | |||
FQ224156.1 | - | 2 | 303 | |||
FQ224523.1 | - | 2 | 306 | |||
FQ224856.1 | - | 2 | 306 | |||
FQ224909.1 | - | 2 | 306 | |||
FQ224921.1 | - | 2 | 303 | |||
FQ224975.1 | - | 2 | 306 |