Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004098.3 | NP_004089.1 | 2, 3 | 1422 | ||
GenBank mRNA | AF301598.1 | - | 2, 3 | 1422 | ||
AK055041.1 | - | 2, 3 | 827 | |||
AK311352.1 | - | 2, 3 | 560 | |||
AL161811.1 | - | 2, 3 | 929 | |||
X68880.1 | - | 2, 3 | 317 |