Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021668.3 | NP_067700.2 | 2 | 428 | ||
XM_006236809.2 | XP_006236871.1 | 329 | ||||
GenBank mRNA | AF185569.1 | - | 370 | |||
BC078836.1 | - | 409 | 7106831 |
Gene Symbol: LOC103690120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236788.2 | XP_006236850.1 | 1 | 329 | ||
XM_008763110.1 | XP_008761332.1 | 2 | 409 |