Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021693.2 | NP_067725.2 | 3, 4 | 352 | ||
GenBank mRNA | AB020480.1 | - | 3, 4 | 352 | ||
AF106937.1 | - | 410 |